Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 3-methyl-3-phenylglycidate, cis + trans, 98%

Click to view available options
Quantity:
50g
250g
1000g
77-83-8
C12H14O3
206.241
MFCD00022338
LQKRYVGRPXFFAV-UHFFFAOYSA-N
ethyl methylphenylglycidate,ethyl 3-methyl-3-phenylglycidate,strawberry aldehyde,fraeseol,empg,c-16 aldehyde,aldehyde c-16,ethyl methylphenyl glycidate,ethyl-3-methyl-3-phenylglycidate,ethyl 2,3-epoxy-3-phenylbutyrate
6501
ethyl 3-methyl-3-phenyloxirane-2-carboxylate
CCOC(=O)C1C(O1)(C)C2=CC=CC=C2
77-83-8
C12H14O3
206.241
MFCD00022338
LQKRYVGRPXFFAV-UHFFFAOYSA-N
ethyl methylphenylglycidate,ethyl 3-methyl-3-phenylglycidate,strawberry aldehyde,fraeseol,empg,c-16 aldehyde,aldehyde c-16,ethyl methylphenyl glycidate,ethyl-3-methyl-3-phenylglycidate,ethyl 2,3-epoxy-3-phenylbutyrate
6501
ethyl 3-methyl-3-phenyloxirane-2-carboxylate
CCOC(=O)C1C(O1)(C)C2=CC=CC=C2
Boiling Point 272°C to 275°C
Quantity 1000g
Flash Point 134°C (273°F)
Beilstein 12299
Formula Weight 206.24
Percent Purity 98%
Chemical Name or Material Ethyl 3-methyl-3-phenylglycidate, cis + trans

RTECSNumber : MW5250000

TSCA : TSCA