Alfa Aesar™ Ethyl 3-(methylthio)propionate, 98%

Manufacturer: Alfa Aesar™ A1174236

Quantity 500g

 View more versions of this product

Catalog No. AAA1174236

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13327-56-5
    Molecular Formula CH3SCH2CH2CO2CH2CH3
    Formula Weight 148.23
    MDL Number MFCD00039900
    Beilstein 1748688
    Synonym 3-(Methylthio)propionic acid ethyl ester; Ethyl 3-(methylmercapto)propionate
    Chemical Name or Material Ethyl 3- (methylthio)propionate

    Specifications

    Formula Weight 148.23
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00039900
    Synonym 3-(Methylthio)propionic acid ethyl ester; Ethyl 3-(methylmercapto)propionate
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Molecular Formula CH3SCH2CH2CO2CH2CH3
    CAS 13327-56-5
    Boiling Point 197°C
    Flash Point 79°C (174°F)
    Beilstein 1748688
    Quantity 500g
    Chemical Name or Material Ethyl 3- (methylthio)propionate
    Assay 98%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit