Alfa Aesar™ Ethyl 3-(methylthio)propionate, 98%

Manufacturer: Alfa Aesar™ A1174214

Quantity 25g

 View more versions of this product

Catalog No. AAA1174214

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 13327-56-5
    Molecular Formula CH3SCH2CH2CO2CH2CH3
    Formula Weight 148.23
    MDL Number MFCD00039900
    Beilstein 1748688
    Synonym 3-(Methylthio)propionic acid ethyl ester; Ethyl 3-(methylmercapto)propionate
    Chemical Name or Material Ethyl 3- (methylthio)propionate

    Specifications

    Formula Weight 148.23
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 13327-56-5
    Beilstein 1748688
    Synonym 3-(Methylthio)propionic acid ethyl ester; Ethyl 3-(methylmercapto)propionate
    Molecular Formula CH3SCH2CH2CO2CH2CH3
    MDL Number MFCD00039900
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Boiling Point 197°C
    Quantity 25g
    Flash Point 79°C (174°F)
    Chemical Name or Material Ethyl 3- (methylthio)propionate
    Assay 98%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit