Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-oxo-3-(2-pyridyl)propionate, 96%, Thermo Scientific™

Catalog Number AAH6453306
Click to view available options
Quantity:
250 mg
1 g
5 g
26510-52-1
C10H11NO3
193.20
MFCD00094022
FQHXWZMJALFSJJ-UHFFFAOYSA-N
ethyl 3-oxo-3-pyridin-2-yl propanoate, ethyl picolinoylacetate, ethyl 3-oxo-3-2-pyridyl propionate, ethyl 3-oxo-3-2-pyridinyl propanoate, ethyl 3-oxo-3-2-pyridyl propanoate, pubchem11088, ethyl-2-pyridoyl acetate, d05hbz, ethyl 3-pyridinyl-3-oxopropionate, 2-3-ethoxy-3-oxopropanoyl pyridine
2736461
CCOC(=O)CC(=O)C1=CC=CC=N1
26510-52-1
C10H11NO3
193.20
MFCD00094022
FQHXWZMJALFSJJ-UHFFFAOYSA-N
ethyl 3-oxo-3-pyridin-2-yl propanoate, ethyl picolinoylacetate, ethyl 3-oxo-3-2-pyridyl propionate, ethyl 3-oxo-3-2-pyridinyl propanoate, ethyl 3-oxo-3-2-pyridyl propanoate, pubchem11088, ethyl-2-pyridoyl acetate, d05hbz, ethyl 3-pyridinyl-3-oxopropionate, 2-3-ethoxy-3-oxopropanoyl pyridine
2736461
CCOC(=O)CC(=O)C1=CC=CC=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.164 g/mL
Boiling Point 100°C to 104°C (0.3 mmHg)
Flash Point >200°C (392°F)
Refractive Index 1.516
Quantity 5 g
Beilstein 145706
Sensitivity Light sensitive
IUPAC Name ethyl 3-oxo-3-pyridin-2-ylpropanoate
Formula Weight 193.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Ethyl 3-oxo-3-(2-pyridyl)propionate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit