Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 3-oxo-3-(2-pyridyl)propionate, 96%

Click to view available options
Quantity:
250mg
1g
5g
26510-52-1
C10H11NO3
193.202
MFCD00094022
FQHXWZMJALFSJJ-UHFFFAOYSA-N
ethyl 3-oxo-3-pyridin-2-yl propanoate,ethyl picolinoylacetate,ethyl 3-oxo-3-2-pyridyl propionate,ethyl 3-oxo-3-2-pyridinyl propanoate,ethyl 3-oxo-3-2-pyridyl propanoate,pubchem11088,ethyl-2-pyridoyl acetate,d05hbz,ethyl 3-pyridinyl-3-oxopropionate,2-3-ethoxy-3-oxopropanoyl pyridine
2736461
ethyl 3-oxo-3-pyridin-2-ylpropanoate
CCOC(=O)CC(=O)C1=CC=CC=N1
26510-52-1
C10H11NO3
193.202
MFCD00094022
FQHXWZMJALFSJJ-UHFFFAOYSA-N
ethyl 3-oxo-3-pyridin-2-yl propanoate,ethyl picolinoylacetate,ethyl 3-oxo-3-2-pyridyl propionate,ethyl 3-oxo-3-2-pyridinyl propanoate,ethyl 3-oxo-3-2-pyridyl propanoate,pubchem11088,ethyl-2-pyridoyl acetate,d05hbz,ethyl 3-pyridinyl-3-oxopropionate,2-3-ethoxy-3-oxopropanoyl pyridine
2736461
ethyl 3-oxo-3-pyridin-2-ylpropanoate
CCOC(=O)CC(=O)C1=CC=CC=N1
Refractive Index 1.5160
Sensitivity Light Sensitive
Density 1.164g/mL
Boiling Point 100°C to 104°C (0.3mmHg)
Quantity 1g
Beilstein 145706
Formula Weight 193.2
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Ethyl 3-oxo-3-(2-pyridyl)propionate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning