Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-oxo-4-(triphenylphosphoranylidene)butyrate, 97%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
13148-05-5
C24H23O3P
390.419
MFCD00192162
QYXSFVCJCUXGJH-UHFFFAOYSA-N
ethyl 3-oxo-4-triphenylphosphoranylidene butyrate,ethyl 3-oxo-4-triphenylphosphoranylidene butanoate,3-ethoxycarbonyl-2-oxopropylidene triphenylphosphorane,ethyl 3-oxo-4-triphenyl-??-phosphanylidene butanoate,acmc-1bsia,ethyl 3-oxo-4-triphenyl,3-ethoxycarbonyl-2-oxopropylidenetriphenyl-phosphorane,3-oxo-4-triphenylphosphoranylidenebutanoic acid ethyl ester
2734128
ethyl 3-oxo-4-(triphenyl-$l^{5}-phosphanylidene)butanoate
CCOC(=O)CC(=O)C=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
13148-05-5
C24H23O3P
390.419
MFCD00192162
QYXSFVCJCUXGJH-UHFFFAOYSA-N
ethyl 3-oxo-4-triphenylphosphoranylidene butyrate,ethyl 3-oxo-4-triphenylphosphoranylidene butanoate,3-ethoxycarbonyl-2-oxopropylidene triphenylphosphorane,ethyl 3-oxo-4-triphenyl-??-phosphanylidene butanoate,acmc-1bsia,ethyl 3-oxo-4-triphenyl,3-ethoxycarbonyl-2-oxopropylidenetriphenyl-phosphorane,3-oxo-4-triphenylphosphoranylidenebutanoic acid ethyl ester
2734128
ethyl 3-oxo-4-(triphenyl-$l^{5}-phosphanylidene)butanoate
CCOC(=O)CC(=O)C=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
Infrared Spectrum Authentic
Melting Point 101°C to 106°C
Packaging Glass bottle
Quantity 5g
Color White to Beige
Assay Percent Range 97%
Linear Formula (C6H5)3P=CHCOCH2CO2C2H5
Formula Weight 390.42
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material Ethyl 3-oxo-4-(triphenylphosphoranylidene)butyrate, 97%
Show More Show Less