Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 3-oxocyclobutanecarboxylate, 97%

Click to view available options
Quantity:
250mg
1g
5g
87121-89-9
C7H10O3
142.154
MFCD09834114
BXBRFSMPBOTZHJ-UHFFFAOYSA-N
ethyl 3-oxocyclobutanecarboxylate,3-oxo-cyclobutanecarboxylic acid ethyl ester,3-oxocyclobutanecarboxylic acid ethyl ester,cyclobutanecarboxylic acid, 3-oxo-, ethyl ester,3-ethoxycarbonylcyclobutanone,ethoxycarbonyl-3-cyclobutanone,ethyl3-oxocyclobutanecarboxylate,3-ethoxycarbonyl-1-cyclobutanone,ethyl 3-oxo-cyclobutanecarboxylate
22467070
ethyl 3-oxocyclobutane-1-carboxylate
CCOC(=O)C1CC(=O)C1
87121-89-9
C7H10O3
142.154
MFCD09834114
BXBRFSMPBOTZHJ-UHFFFAOYSA-N
ethyl 3-oxocyclobutanecarboxylate,3-oxo-cyclobutanecarboxylic acid ethyl ester,3-oxocyclobutanecarboxylic acid ethyl ester,cyclobutanecarboxylic acid, 3-oxo-, ethyl ester,3-ethoxycarbonylcyclobutanone,ethoxycarbonyl-3-cyclobutanone,ethyl3-oxocyclobutanecarboxylate,3-ethoxycarbonyl-1-cyclobutanone,ethyl 3-oxo-cyclobutanecarboxylate
22467070
ethyl 3-oxocyclobutane-1-carboxylate
CCOC(=O)C1CC(=O)C1
Applications
Ethyl 3-oxocyclobutanecarboxylate is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Quantity 1g
Formula Weight 142.15
Percent Purity 97%
Chemical Name or Material Ethyl 3-oxocyclobutanecarboxylate

TSCA : No

Recommended Storage : Ambient temperatures