Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Ethyl 3-phenylglycidate, cis + trans, 90+%
Click to view available options
Quantity:
50mL
250mL
1000mL
Specifications
Sensitivity | Moisture sensitive |
Density | 1.102 |
Refractive Index | 1.518 |
Flash Point | >110°C (230°F) |
Quantity | 50mL |
Boiling Point | 95°C to 97°C (0.5mmHg) |
Formula Weight | 192.21 |
Percent Purity | ≥90% |
Chemical Name or Material | Ethyl 3-phenylglycidate, cis + trans |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
EINECSNumber : 204-467-3
TSCA : Yes
Recommended Storage : Keep cold