Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-phenylpropionate, 98%, Acros Organics

Click to view available options
Quantity:
100g
2021-28-5
C11H14O2
178.231
MFCD00009206
JAGZUIGGHGTFHO-UHFFFAOYSA-N
ethyl 3-phenylpropionate,ethyl hydrocinnamate,ethyl dihydrocinnamate,benzenepropanoic acid, ethyl ester,3-phenylpropionic acid ethyl ester,hydrocinnamic acid, ethyl ester,ethylhydrocinnamoate,ethyl benzenepropanoate,unii-58e9z9v64d,ethylphenyl propanoate
16237
ethyl 3-phenylpropanoate
CCOC(=O)CCC1=CC=CC=C1
2021-28-5
C11H14O2
178.231
MFCD00009206
JAGZUIGGHGTFHO-UHFFFAOYSA-N
ethyl 3-phenylpropionate,ethyl hydrocinnamate,ethyl dihydrocinnamate,benzenepropanoic acid, ethyl ester,3-phenylpropionic acid ethyl ester,hydrocinnamic acid, ethyl ester,ethylhydrocinnamoate,ethyl benzenepropanoate,unii-58e9z9v64d,ethylphenyl propanoate
16237
ethyl 3-phenylpropanoate
CCOC(=O)CCC1=CC=CC=C1
Density 1.1g/mL
Refractive Index 1.4940 to 1.4960 (20°C, 589nm)
Infrared Spectrum Authentic
Quantity 100g
Color Colorless to Yellow
Assay Percent Range 98%
Formula Weight 178.23
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Ethyl 3-phenylpropionate, 98%