Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-pyridylacetate 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
39931-77-6
C9H11NO2
165.19
MFCD00006411
RPWXYCRIAGBAGY-UHFFFAOYSA-N
ethyl 3-pyridylacetate,ethyl 2-pyridin-3-yl acetate,ethyl 3-pyridineacetate,3-pyridineacetic acid, ethyl ester,acetic acid, 2-3-pyridyl-, ethyl ester,pyridin-3-yl-acetic acid ethyl ester,3-pyridylacetic acid ethyl ester,ethyl 2-3-pyridyl acetate,pubchem20506,ethyl-3-pyridyl-acetate
96490
ethyl 2-pyridin-3-ylacetate
CCOC(=O)CC1=CN=CC=C1
39931-77-6
C9H11NO2
165.19
MFCD00006411
RPWXYCRIAGBAGY-UHFFFAOYSA-N
ethyl 3-pyridylacetate,ethyl 2-pyridin-3-yl acetate,ethyl 3-pyridineacetate,3-pyridineacetic acid, ethyl ester,acetic acid, 2-3-pyridyl-, ethyl ester,pyridin-3-yl-acetic acid ethyl ester,3-pyridylacetic acid ethyl ester,ethyl 2-3-pyridyl acetate,pubchem20506,ethyl-3-pyridyl-acetate
96490
ethyl 2-pyridin-3-ylacetate
CCOC(=O)CC1=CN=CC=C1
Color Colorless to Yellow
Refractive Index 1.4990 to 1.501
Density 1.0800g/mL
Boiling Point 80°C (0.4mmHg)
Flash Point >110°C
Specific Gravity 1.08
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula (C5H4N)CH2COOC2H5
Formula Weight 165.19
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Ethyl 3-pyridylacetate, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 254-707-6

RTECSNumber : UR8950000