Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-(trimethylsilyl)propiolate, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass Bottle
16205-84-8
C8H14O2Si
170.283
MFCD00074886
QRBKPHYAIRLCLK-UHFFFAOYSA-N
ethyl 3-trimethylsilyl propiolate,ethyl 3-trimethylsilyl propynoate,ethyl 3-trimethylsilyl prop-2-ynoate,2-propynoic acid, 3-trimethylsilyl-, ethyl ester,ethyl 3-trimethylsilyl-2-propynoate,acmc-1bvr4,ethyl 3-trimethylsily propynoate,ethyl3-trimethylsilyl propiolate,ethyl 3-trimethylsilyl propalate,trimethylsilylpropiolic acid ethyl ester
568601
ethyl 3-trimethylsilylprop-2-ynoate
CCOC(=O)C#C[Si](C)(C)C
16205-84-8
C8H14O2Si
170.283
MFCD00074886
QRBKPHYAIRLCLK-UHFFFAOYSA-N
ethyl 3-trimethylsilyl propiolate,ethyl 3-trimethylsilyl propynoate,ethyl 3-trimethylsilyl prop-2-ynoate,2-propynoic acid, 3-trimethylsilyl-, ethyl ester,ethyl 3-trimethylsilyl-2-propynoate,acmc-1bvr4,ethyl 3-trimethylsily propynoate,ethyl3-trimethylsilyl propiolate,ethyl 3-trimethylsilyl propalate,trimethylsilylpropiolic acid ethyl ester
568601
ethyl 3-trimethylsilylprop-2-ynoate
CCOC(=O)C#C[Si](C)(C)C
Packaging Glass Bottle
Quantity 5g
Infrared Spectrum Authentic
Color Colorless
Density 0.8970g/mL
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Assay Percent Range 99%
Linear Formula (CH3)3SiCidentical toCCOOCH2CH3
Formula Weight 170.28
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Ethyl 3-(trimethylsilyl)propiolate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning