Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 97+%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
195062-62-5
C15H21BO4
276.139
MFCD03453659
NCVIYKCFTYSAGN-UHFFFAOYSA-N
ethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate,4-ethoxycarbonylphenylboronic acid pinacol ester,4-ethoxycarbonylphenylboronic acid, pinacol ester,4-carboethoxyphenylboronic acid pinacol ester,ethyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate,ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate,4-ethoxycarbonyl benzeneboronic acid pinacol ester,2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester
2734618
ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
195062-62-5
C15H21BO4
276.139
MFCD03453659
NCVIYKCFTYSAGN-UHFFFAOYSA-N
ethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate,4-ethoxycarbonylphenylboronic acid pinacol ester,4-ethoxycarbonylphenylboronic acid, pinacol ester,4-carboethoxyphenylboronic acid pinacol ester,ethyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate,ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate,4-ethoxycarbonyl benzeneboronic acid pinacol ester,2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester
2734618
ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
Quantity 5g
Infrared Spectrum Authentic
Density 1.0520g/mL
Refractive Index 1.4980 to 1.5000 (20°C, 589nm)
Color Yellow
Packaging Glass bottle
Assay Percent Range 97+%
Formula Weight 276.14
Physical Form Viscous Liquid
Percent Purity ≥97.0%
Chemical Name or Material Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Show More Show Less

Recommended Storage : Refrigerator +4°C