Learn More
Ethyl 4,4,4-trifluoro-2-butynoate, 97%, Thermo Scientific Chemicals
Description
Ethyl 4,4,4-trifluoro-2-butynoate is the suitable reagent used to investigate the regioselectivity of the insertion reaction with cyclometalated iridium and rhodium complexes. It may be used in the preparation of ethyl (Z)-3-iodo-4,4,4-trifluoro-2-butenoate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl 4,4,4-trifluoro-2-butynoate is the suitable reagent used to investigate the regioselectivity of the insertion reaction with cyclometalated iridium and rhodium complexes. It may be used in the preparation of ethyl (Z)-3-iodo-4,4,4-trifluoro-2-butenoate.
Solubility
Soluble in water (partly).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Density | 1.162 |
Boiling Point | 96°C to 98°C |
Flash Point | 6°C (43°F) |
Refractive Index | 1.35 |
Linear Formula | CF3C≡CCO2CH2CH3 |
Quantity | 5 g |
UN Number | UN3272 |
Beilstein | 3539414 |
Synonym | ethyl 4,4,4-trifluoro-2-butynoate, 4,4,4-trifluoro-2-butynoic acid ethyl ester, 4,4,4-trifluoro-but-2-ynoic acid ethyl ester, pubchem10880, acmc-1bc5r, ethyl4,4,4-trifluoro-2-butynoate, ethyl 4,4,4-trifluoro-2-butynate, trifluoro-2-butynoic acid ethyl ester, ethyl 4,4,4-trifluorobutynoate, ethyl 4,4,4-tris fluoranyl but-2-ynoate |
Solubility Information | Soluble in water (partly). |
Show More |
Safety and Handling
Hazard Category | H225-H302+H312-H315-H319-H335 |
Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.