Alfa Aesar™ Ethyl 4,4-difluoroacetoacetate, 98%

Manufacturer: Alfa Aesar™ H3192203

Quantity 1g

 View more versions of this product

Catalog No. AAH3192203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 352-24-9
    Molecular Formula C6H8F2O3
    Formula Weight 166.13
    MDL Number MFCD00039277
    UN Number UN1993
    Chemical Name or Material Ethyl 4,4-difluoroacetoacetate

    Specifications

    Formula Weight 166.13
    TSCA TSCA
    Molecular Formula C6H8F2O3
    MDL Number MFCD00039277
    UN Number UN1993
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    CAS 352-24-9
    Chemical Name or Material Ethyl 4,4-difluoroacetoacetate
    Assay 98%

    GHS H Statement
    H226-H315-H319
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit