Alfa Aesar™ Ethyl 4,4-difluorocyclohexanecarboxylate, 97%

Manufacturer: Alfa Aesar™ H2575406

Quantity 5g

 View more versions of this product

Catalog No. AAH2575406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 178312-47-5
    Molecular Formula C9H14F2O2
    Formula Weight 192.21
    MDL Number MFCD03788494
    Synonym 4,4-Difluorocyclohexanecarboxylic acid ethyl ester
    Chemical Name or Material Ethyl 4,4-difluorocyclohexanecarboxylate

    Specifications

    Formula Weight 192.21
    Molecular Formula C9H14F2O2
    MDL Number MFCD03788494
    Boiling Point 84° to 86°C (8mmHg)
    Quantity 5g
    CAS 178312-47-5
    Synonym 4,4-Difluorocyclohexanecarboxylic acid ethyl ester
    Chemical Name or Material Ethyl 4,4-difluorocyclohexanecarboxylate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit