Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-acetyl-5-oxohexanoate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
2832-10-2
C10H16O4
200.23
MFCD00009211
CVPCFJRJUCNUJG-VQHVLOKHSA-N
ethyl 4,4-diacetylbutyrate,4-acetyl-5-oxohexanoic acid ethyl ester,4,4-diacetylbutyric acid ethyl ester,ethyl 4-acetyl-5-oxohexanoate,4-acetyl-5-oxo-hexanoic acid ethyl ester,hexanoic acid,4-acetyl-5-oxo-, ethyl ester,hexanoic acid, 4-acetyl-5-oxo-, ethyl ester
2733934
ethyl 4-acetyl-5-oxohexanoate
CCOC(=O)CCC(C(=O)C)C(=O)C
2832-10-2
C10H16O4
200.23
MFCD00009211
CVPCFJRJUCNUJG-VQHVLOKHSA-N
ethyl 4,4-diacetylbutyrate,4-acetyl-5-oxohexanoic acid ethyl ester,4,4-diacetylbutyric acid ethyl ester,ethyl 4-acetyl-5-oxohexanoate,4-acetyl-5-oxo-hexanoic acid ethyl ester,hexanoic acid,4-acetyl-5-oxo-, ethyl ester,hexanoic acid, 4-acetyl-5-oxo-, ethyl ester
2733934
ethyl 4-acetyl-5-oxohexanoate
CCOC(=O)CCC(C(=O)C)C(=O)C
Refractive Index 1.4530 to 1.4580 (20°C, 589nm)
Quantity 5g
Infrared Spectrum Authentic
Color Yellow
Density 1.067g/mL
NMR 97.5% min.
Assay Percent Range 98%
Beilstein 03, IV, 1786
Formula Weight 200.23
Physical Form Liquid
Chemical Name or Material Ethyl 4-acetyl-5-oxohexanoate
Show More Show Less