Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Acetyl-5-oxohexanoate 96.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
2832-10-2
C10H16O4
200.23
MFCD00009211
CVPCFJRJUCNUJG-VQHVLOKHSA-N
ethyl 4,4-diacetylbutyrate,4-acetyl-5-oxohexanoic acid ethyl ester,4,4-diacetylbutyric acid ethyl ester,ethyl 4-acetyl-5-oxohexanoate,4-acetyl-5-oxo-hexanoic acid ethyl ester,hexanoic acid,4-acetyl-5-oxo-, ethyl ester,hexanoic acid, 4-acetyl-5-oxo-, ethyl ester
2733934
ethyl 4-acetyl-5-oxohexanoate
CCOC(=O)CCC(C(=O)C)C(=O)C
2832-10-2
C10H16O4
200.23
MFCD00009211
CVPCFJRJUCNUJG-VQHVLOKHSA-N
ethyl 4,4-diacetylbutyrate,4-acetyl-5-oxohexanoic acid ethyl ester,4,4-diacetylbutyric acid ethyl ester,ethyl 4-acetyl-5-oxohexanoate,4-acetyl-5-oxo-hexanoic acid ethyl ester,hexanoic acid,4-acetyl-5-oxo-, ethyl ester,hexanoic acid, 4-acetyl-5-oxo-, ethyl ester
2733934
ethyl 4-acetyl-5-oxohexanoate
CCOC(=O)CCC(C(=O)C)C(=O)C
Boiling Point 182°C
Color Yellow
Quantity 25g
Formula Weight 200.23
Physical Form Clear Liquid at 20°C
Percent Purity ≥96.0% (GC)
Chemical Name or Material Ethyl 4-Acetyl-5-oxohexanoate