Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-amino-1-piperidinecarboxylate, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
58859-46-4
C8H16N2O2
172.228
MFCD00006484
GQQQULCEHJQUJT-UHFFFAOYSA-N
ethyl 4-amino-1-piperidinecarboxylate,4-amino-1-carbethoxypiperidine,unii-54815srp4z,1-piperidinecarboxylic acid, 4-amino-, ethyl ester,ethyl 4-aminopiperidinecarboxylate,ethyl-4-amino-1-piperidine carboxylate,4-amino-1-piperidinecarboxylic acid ethyl ester,ethyl-4-aminopiperidin-1-carboxylat,pubchem2064,acmc-209m7k
100867
ethyl 4-aminopiperidine-1-carboxylate
CCOC(=O)N1CCC(CC1)N
58859-46-4
C8H16N2O2
172.228
MFCD00006484
GQQQULCEHJQUJT-UHFFFAOYSA-N
ethyl 4-amino-1-piperidinecarboxylate,4-amino-1-carbethoxypiperidine,unii-54815srp4z,1-piperidinecarboxylic acid, 4-amino-, ethyl ester,ethyl 4-aminopiperidinecarboxylate,ethyl-4-amino-1-piperidine carboxylate,4-amino-1-piperidinecarboxylic acid ethyl ester,ethyl-4-aminopiperidin-1-carboxylat,pubchem2064,acmc-209m7k
100867
ethyl 4-aminopiperidine-1-carboxylate
CCOC(=O)N1CCC(CC1)N
Refractive Index 1.4810 to 1.4830 (20°C, 589nm)
Packaging Glass bottle
Density 1.0040g/mL
Quantity 5g
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Formula Weight 172.23
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material Ethyl 4-amino-1-piperidinecarboxylate, 95%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE