Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-aminobenzoate, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
94-09-7
C9H11NO2
165.19
BLFLLBZGZJTVJG-UHFFFAOYSA-N
benzocaine,ethyl aminobenzoate,ethyl p-aminobenzoate,americaine,anesthesin,anaesthesin,ethoform,norcaine,orthesin,parathesin
2337
CHEBI:116735
ethyl 4-aminobenzoate
CCOC(=O)C1=CC=C(C=C1)N
94-09-7
C9H11NO2
165.19
BLFLLBZGZJTVJG-UHFFFAOYSA-N
benzocaine,ethyl aminobenzoate,ethyl p-aminobenzoate,americaine,anesthesin,anaesthesin,ethoform,norcaine,orthesin,parathesin
2337
CHEBI:116735
ethyl 4-aminobenzoate
CCOC(=O)C1=CC=C(C=C1)N
Boiling Point 172°C (17.0 hPa)
Flash Point >100°C
Packaging Plastic bottle
Melting Point 89.0°C to 92.0°C
Color White
Quantity 2.5kg
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula H2NC6H4CO2C2H5
Beilstein 14,422
Merck Index 15,1088
Solubility Information Solubility in water: 1g/2500mL. Other solubilities: 1g/5mL alcohol,1g/2mL chloroform,1g/4mL ether,ca. 1g/40mL expressed almond oil or olive oil
Formula Weight 165.19
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Ethyl 4-aminobenzoate
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning