Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-bromobutyrate, 97+%, Thermo Scientific Chemicals

Catalog Number AC173351000
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
2969-81-5
C6H11BrO2
195.06
XBPOBCXHALHJFP-UHFFFAOYSA-N
ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl ester
76300
ethyl 4-bromobutanoate
CCOC(=O)CCCBr
2969-81-5
C6H11BrO2
195.06
XBPOBCXHALHJFP-UHFFFAOYSA-N
ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl ester
76300
ethyl 4-bromobutanoate
CCOC(=O)CCCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.3600g/mL
Boiling Point 80.0°C to 82.0°C (10.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4549 to 1.4569
Linear Formula Br(CH2)3CO2C2H5
Quantity 100 mL
Beilstein 02,283
Specific Gravity 1.36
Solubility Information Solubility in water: insoluble.
Formula Weight 195.06
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Ethyl 4-bromobutyrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
EINECSNumber 221-005-6

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit