Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 4-chloro-3-nitrobenzoate, 97%

Click to view available options
Quantity:
5g
25g
16588-16-2
C9H8ClNO4
229.616
MFCD00219621
BLNLZRQIUGDTAO-UHFFFAOYSA-N
benzoic acid, 4-chloro-3-nitro-, ethyl ester,4-chloro-3-nitrobenzoic acid ethyl ester,4-chloro-3-nitro-benzoic acid ethyl ester,ethyl 4-chloranyl-3-nitro-benzoate,acmc-20a7tu,ethyl4-chloro-3-nitrobenzoate,ethyl 4-chloro-3-nitro-benzoate,ethyl-4-chloro-3-nitro-benzoate,ethyl-4-chloro-3-nitrobenzoate,3-nitro-4-chlorobenzoic acid ethyl ester
1268247
ethyl 4-chloro-3-nitrobenzoate
CCOC(=O)C1=CC(=C(C=C1)Cl)[N+](=O)[O-]
16588-16-2
C9H8ClNO4
229.616
MFCD00219621
BLNLZRQIUGDTAO-UHFFFAOYSA-N
benzoic acid, 4-chloro-3-nitro-, ethyl ester,4-chloro-3-nitrobenzoic acid ethyl ester,4-chloro-3-nitro-benzoic acid ethyl ester,ethyl 4-chloranyl-3-nitro-benzoate,acmc-20a7tu,ethyl4-chloro-3-nitrobenzoate,ethyl 4-chloro-3-nitro-benzoate,ethyl-4-chloro-3-nitro-benzoate,ethyl-4-chloro-3-nitrobenzoate,3-nitro-4-chlorobenzoic acid ethyl ester
1268247
ethyl 4-chloro-3-nitrobenzoate
CCOC(=O)C1=CC(=C(C=C1)Cl)[N+](=O)[O-]
Melting Point 53°C to 55°C
Quantity 5g
Formula Weight 229.62
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloro-3-nitrobenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures