Alfa Aesar™ Ethyl 4-chloro-6,8-difluoroquinoline-3-carboxylate

Manufacturer: Alfa Aesar™ H5050303

Quantity 1g

 View more versions of this product

Catalog No. AAH5050303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 150258-20-1
    Molecular Formula C12H8ClF2NO2
    Formula Weight 271.65
    MDL Number MFCD00173353
    Synonym 4-Chloro-6,8-difluoroquinoline-3-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl 4-chloro-6,8-difluoroquinoline-3-carboxylate

    Specifications

    Formula Weight 271.65
    MDL Number MFCD00173353
    Quantity 1g
    Molecular Formula C12H8ClF2NO2
    CAS 150258-20-1
    Synonym 4-Chloro-6,8-difluoroquinoline-3-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl 4-chloro-6,8-difluoroquinoline-3-carboxylate

    Description

    Applications
    Ethyl 4-chloro-6,8-difluoroquinoline-3-carboxylate is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit