Alfa Aesar™ Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate, 97%

Manufacturer: Alfa Aesar™ H50515MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50515MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22931-71-1
    Molecular Formula C13H12ClNO3
    Formula Weight 265.7
    MDL Number MFCD00173417
    Synonym 4-Chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate

    Specifications

    Formula Weight 265.7
    Molecular Formula C13H12ClNO3
    MDL Number MFCD00173417
    CAS 22931-71-1
    Quantity 250mg
    Synonym 4-Chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester
    Melting Point 88° to 90°C
    Chemical Name or Material Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate
    Assay 97%

    Description

    Applications
    Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

    Notes
    Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit