Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H5051503

Quantity 1g

 View more versions of this product

Catalog No. AAH5051503


  Check Availability
Add to cart

Specifications

Description

Applications
Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Description & Specifications

Chemical Identifiers

22931-71-1
265.7
4-Chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester
C13H12ClNO3
MFCD00173417
Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate

Specifications

22931-71-1
265.7
4-Chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester
1g
97%
C13H12ClNO3
MFCD00173417
88° to 90°C
Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit