Alfa Aesar™ Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate, 97%

Manufacturer: Alfa Aesar™ H5051306

Quantity 5g

 View more versions of this product

Catalog No. AAH5051306

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 77173-67-2
    Molecular Formula C13H9ClN2O2
    Formula Weight 260.68
    MDL Number MFCD00173414
    Synonym 4-Chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate

    Specifications

    Formula Weight 260.68
    Molecular Formula C13H9ClN2O2
    MDL Number MFCD00173414
    CAS 77173-67-2
    Synonym 4-Chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    Chemical Name or Material Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
    Assay 97%

    GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit