Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
77173-67-2
C13H9ClN2O2
260.68
MHOFAUCLSGVKOH-UHFFFAOYSA-N
ethyl4-chloro-8-cyanoquinoline-3-carboxylate,ethyl 4-chloro-8-cyano-3-quinolinecarboxylate,pubchem19877,4-chloro-8-cyanoquinoline-3-carboxylicacidethyleste,4-chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester,ethyl 4-chloro-8-cyanoquinoxaline-3-carboxylate
3761179
ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
CCOC(=O)C1=C(C2=CC=CC(=C2N=C1)C#N)Cl
77173-67-2
C13H9ClN2O2
260.68
MHOFAUCLSGVKOH-UHFFFAOYSA-N
ethyl4-chloro-8-cyanoquinoline-3-carboxylate,ethyl 4-chloro-8-cyano-3-quinolinecarboxylate,pubchem19877,4-chloro-8-cyanoquinoline-3-carboxylicacidethyleste,4-chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester,ethyl 4-chloro-8-cyanoquinoxaline-3-carboxylate
3761179
ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
CCOC(=O)C1=C(C2=CC=CC(=C2N=C1)C#N)Cl
Packaging Glass Bottle
Water 1% max.
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Formula Weight 260.68
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning