Learn More
Alfa Aesar™ Ethyl 4-chloro-8-methoxyquinoline-3-carboxylate
Manufacturer:  Alfa Aesar™ H50509MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH50509MD
  Check Availability
Add to cart
Specifications
Description
Applications
Ethyl 4-chloro-8-methoxyquinoline-3-carboxylate is used as medical intermediates.
Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.
Description & Specifications
Chemical Identifiers
27568-05-4
265.7
4-Chloro-8-methoxyquinoline-3-carboxylic acid ethyl ester
C13H12ClNO3
MFCD00173395
Ethyl 4-chloro-8-methoxyquinoline-3-carboxylate
Specifications
27568-05-4
265.7
4-Chloro-8-methoxyquinoline-3-carboxylic acid ethyl ester
Ethyl 4-chloro-8-methoxyquinoline-3-carboxylate
C13H12ClNO3
MFCD00173395
250mg