Alfa Aesar™ Ethyl 4-chloro-8-methylquinoline-3-carboxylate

Manufacturer: Alfa Aesar™ H50510MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50510MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 37041-32-0
    Molecular Formula C13H12ClNO2
    Formula Weight 249.7
    MDL Number MFCD00173399
    Synonym 4-Chloro-8-methylquinoline-3-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl 4-chloro-8-methylquinoline-3-carboxylate

    Specifications

    Formula Weight 249.7
    Molecular Formula C13H12ClNO2
    MDL Number MFCD00173399
    CAS 37041-32-0
    Synonym 4-Chloro-8-methylquinoline-3-carboxylic acid ethyl ester
    Quantity 250mg
    Chemical Name or Material Ethyl 4-chloro-8-methylquinoline-3-carboxylate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit