Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 4-chloroacetoacetate, 97%

Click to view available options
Quantity:
50g
250g
1000g
638-07-3
C6H9ClO3
164.585
MFCD00000939
OHLRLMWUFVDREV-UHFFFAOYSA-N
ethyl 4-chloroacetoacetate,4-chloroacetoacetic acid ethyl ester,butanoic acid, 4-chloro-3-oxo-, ethyl ester,ethyl gamma-chloroacetoacetate,unii-5ae82250cm,ccris 6791,ethyl chloroacetyl acetate,butanoic acid, 4-chloro-3-oxo-ethylester,ethyl 4-chloro-3-oxobutyrate,ethyl 4-chloro-3-oxo-butanoate
69484
ethyl 4-chloro-3-oxobutanoate
CCOC(=O)CC(=O)CCl
638-07-3
C6H9ClO3
164.585
MFCD00000939
OHLRLMWUFVDREV-UHFFFAOYSA-N
ethyl 4-chloroacetoacetate,4-chloroacetoacetic acid ethyl ester,butanoic acid, 4-chloro-3-oxo-, ethyl ester,ethyl gamma-chloroacetoacetate,unii-5ae82250cm,ccris 6791,ethyl chloroacetyl acetate,butanoic acid, 4-chloro-3-oxo-ethylester,ethyl 4-chloro-3-oxobutyrate,ethyl 4-chloro-3-oxo-butanoate
69484
ethyl 4-chloro-3-oxobutanoate
CCOC(=O)CC(=O)CCl
Applications
Ethyl 4-chloroacetoacetate is used as a precursor in the preparation of phosphorous ylides. It finds application in regular film coating, pharmaceutical coating and formulation.

Solubility
Miscible with water and most organic solvents.

Notes
Incompatible with strong acids, strong bases, reducing agents and strong oxidizing agents. Store in a cool place.
Density 1.212
Refractive Index 1.452
Melting Point -8°C
Boiling Point 220°C
Quantity 250g
Flash Point 92°C (198°F)
UN Number UN2922
Beilstein 1761275
Solubility Information Miscible with water and most organic solvents.
Formula Weight 164.59
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloroacetoacetate
Show More Show Less

GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311-H314-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 211-317-0

RTECSNumber : AK5110000

TSCA : Yes

Recommended Storage : Ambient temperatures