Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 4-fluorobenzoate, 99%

Click to view available options
Quantity:
5g
25g
100g
451-46-7
C9H9FO2
168.167
MFCD00000351
UMPRJGKLMUDRHL-UHFFFAOYSA-N
4-fluorobenzoic acid ethyl ester,ethyl p-fluorobenzoate,benzoic acid, 4-fluoro-, ethyl ester,p-fluorobenzoic acid, ethyl ester,ethyl4-fluorobenzoate,4-fluoro-benzoic acid ethyl ester,benzoic acid, p-fluoro-, ethyl ester,ethyep-fluorobenzoate,pubchem3471,ethyl-4-fluorobenzoate
67976
ethyl 4-fluorobenzoate
CCOC(=O)C1=CC=C(C=C1)F
451-46-7
C9H9FO2
168.167
MFCD00000351
UMPRJGKLMUDRHL-UHFFFAOYSA-N
4-fluorobenzoic acid ethyl ester,ethyl p-fluorobenzoate,benzoic acid, 4-fluoro-, ethyl ester,p-fluorobenzoic acid, ethyl ester,ethyl4-fluorobenzoate,4-fluoro-benzoic acid ethyl ester,benzoic acid, p-fluoro-, ethyl ester,ethyep-fluorobenzoate,pubchem3471,ethyl-4-fluorobenzoate
67976
ethyl 4-fluorobenzoate
CCOC(=O)C1=CC=C(C=C1)F
Flash Point 81°C (177°F)
Melting Point 25°C to 27°C
Boiling Point 209°C to 210°C
Quantity 5g
Odor Recognizable
Beilstein 1866794
Merck Index 14,4171
Formula Weight 168.17
Percent Purity 99%
Chemical Name or Material Ethyl 4-fluorobenzoate