Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-fluorobenzoate, 99%, Thermo Scientific Chemicals

Catalog Number AAB2148414
Click to view available options
Quantity:
5 g
25 g
451-46-7
C9H9FO2
168.167
MFCD00000351
UMPRJGKLMUDRHL-UHFFFAOYSA-N
4-fluorobenzoic acid ethyl ester, ethyl p-fluorobenzoate, benzoic acid, 4-fluoro-, ethyl ester, p-fluorobenzoic acid, ethyl ester, ethyl4-fluorobenzoate, 4-fluoro-benzoic acid ethyl ester, benzoic acid, p-fluoro-, ethyl ester, ethyep-fluorobenzoate, pubchem3471, ethyl-4-fluorobenzoate
67976
ethyl 4-fluorobenzoate
CCOC(=O)C1=CC=C(C=C1)F
451-46-7
C9H9FO2
168.167
MFCD00000351
UMPRJGKLMUDRHL-UHFFFAOYSA-N
4-fluorobenzoic acid ethyl ester, ethyl p-fluorobenzoate, benzoic acid, 4-fluoro-, ethyl ester, p-fluorobenzoic acid, ethyl ester, ethyl4-fluorobenzoate, 4-fluoro-benzoic acid ethyl ester, benzoic acid, p-fluoro-, ethyl ester, ethyep-fluorobenzoate, pubchem3471, ethyl-4-fluorobenzoate
67976
ethyl 4-fluorobenzoate
CCOC(=O)C1=CC=C(C=C1)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 25°C to 27°C
Density 1.146
Boiling Point 209°C to 210°C
Flash Point 81°C (177°F)
Odor Recognizable
Refractive Index 1.487
Quantity 25 g
Beilstein 1866794
Merck Index 14,4171
Formula Weight 168.17
Percent Purity 99%
Chemical Name or Material Ethyl 4-fluorobenzoate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-194-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit