Alfa Aesar™ Ethyl (4-formyl-2-methoxyphenoxy)acetate

Manufacturer: Alfa Aesar™ H5049103

Quantity 1g

 View more versions of this product

Catalog No. AAH5049103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 51264-70-1
    Molecular Formula C12H14O5
    Formula Weight 238.24
    MDL Number MFCD01764358
    Synonym (4-Formyl-2-methoxyphenoxy)acetic acid ethyl ester
    Chemical Name or Material Ethyl (4-formyl-2-methoxyphenoxy)acetate

    Specifications

    Formula Weight 238.24
    Molecular Formula C12H14O5
    MDL Number MFCD01764358
    CAS 51264-70-1
    Synonym (4-Formyl-2-methoxyphenoxy)acetic acid ethyl ester
    Quantity 1g
    Chemical Name or Material Ethyl (4-formyl-2-methoxyphenoxy)acetate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit