Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 4-hydroxy-2-methylpyrimidine-5-carboxylate, 97%

Click to view available options
Quantity:
1g
5g
53135-24-3
C8H10N2O3
182.179
MFCD00223606
KTZQDIINDVWLES-UHFFFAOYSA-N
ethyl 4-hydroxy-2-methylpyrimidine-5-carboxylate,ethyl 2-methyl-6-oxo-1,6-dihydropyrimidine-5-carboxylate,4-hydroxy-2-methyl-pyrimidine-5-carboxylic acid ethyl ester,4-hydroxy-2-methylpyrimidine-5-carboxylic acidethyl ester,ethyl4-hydroxy-2-methylpyrimidine-5-carboxylate,ethyl 4-hydroxy-2-methyl-pyrimidine-5-carboxylate,4-hydroxy-2-methylpyrimidine-5-carboxylic acid ethyl ester,ethyl 4-hydroxy-2-methyl-5-pyrimidine carboxylate,4-hydroxy-2-methyl-pyrimidine-5-carboxylic acidethyl ester,5-pyrimidinecarboxylic acid, 4-hydroxy-2-methyl-, ethyl ester
293750
ethyl 2-methyl-6-oxo-1H-pyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(NC1=O)C
53135-24-3
C8H10N2O3
182.179
MFCD00223606
KTZQDIINDVWLES-UHFFFAOYSA-N
ethyl 4-hydroxy-2-methylpyrimidine-5-carboxylate,ethyl 2-methyl-6-oxo-1,6-dihydropyrimidine-5-carboxylate,4-hydroxy-2-methyl-pyrimidine-5-carboxylic acid ethyl ester,4-hydroxy-2-methylpyrimidine-5-carboxylic acidethyl ester,ethyl4-hydroxy-2-methylpyrimidine-5-carboxylate,ethyl 4-hydroxy-2-methyl-pyrimidine-5-carboxylate,4-hydroxy-2-methylpyrimidine-5-carboxylic acid ethyl ester,ethyl 4-hydroxy-2-methyl-5-pyrimidine carboxylate,4-hydroxy-2-methyl-pyrimidine-5-carboxylic acidethyl ester,5-pyrimidinecarboxylic acid, 4-hydroxy-2-methyl-, ethyl ester
293750
ethyl 2-methyl-6-oxo-1H-pyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(NC1=O)C
Quantity 1g
Beilstein 152549
Formula Weight 182.18
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Ethyl 4-hydroxy-2-methylpyrimidine-5-carboxylate