Alfa Aesar™ Ethyl 4-hydroxypiperidine-1-carboxylate, 98%

Manufacturer: Alfa Aesar™ H3138806

Quantity 5g

 View more versions of this product

Catalog No. AAH3138806

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 65214-82-6
    Molecular Formula C8H15NO3
    Formula Weight 173.21
    MDL Number MFCD00086880
    Synonym 4-Hydroxypiperidine-1-carboxylic acid ethyl ester; 1-Ethoxycarbonyl-1-piperidinol
    Chemical Name or Material Ethyl 4-hydroxypiperidine-1-carboxylate

    Specifications

    Formula Weight 173.21
    Molecular Formula C8H15NO3
    MDL Number MFCD00086880
    CAS 65214-82-6
    Synonym 4-Hydroxypiperidine-1-carboxylic acid ethyl ester; 1-Ethoxycarbonyl-1-piperidinol
    Quantity 5g
    Chemical Name or Material Ethyl 4-hydroxypiperidine-1-carboxylate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit