Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 4-hydroxypiperidine-1-carboxylate, 98%

Manufacturer:  Alfa Aesar™ H3138822

Quantity 100g

 View more versions of this product

Catalog No. AAH3138822

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

65214-82-6
173.21
4-Hydroxypiperidine-1-carboxylic acid ethyl ester; 1-Ethoxycarbonyl-1-piperidinol
C8H15NO3
MFCD00086880
Ethyl 4-hydroxypiperidine-1-carboxylate

Specifications

65214-82-6
173.21
4-Hydroxypiperidine-1-carboxylic acid ethyl ester; 1-Ethoxycarbonyl-1-piperidinol
Ethyl 4-hydroxypiperidine-1-carboxylate
C8H15NO3
MFCD00086880
100g
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit