Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 4-isopropylpiperazine-1-carboxylate, 98%

Click to view available options
Quantity:
1g
5g
61014-91-3
C10H20N2O2
200.282
MFCD08064222
HRMZRPQIFGCXML-UHFFFAOYSA-N
ethyl 4-1-methylethyl piperazine-1-carboxylate,ethyl 4-isopropylpiperazine-1-carboxylate,ethyl 4-propan-2-yl piperazine-1-carboxylate,acmc-20akz8,ethyl 4-methylethyl piperazinecarboxylate,ethyl 4-isopropyl-1-piperazinecarboxylate,ethyl 4-1-methylethyl-1-piperazinylcarboxylate,4-isopropylpiperazine-1-carboxylic acid ethyl ester,1-piperazinecarboxylic acid, 4-1-methylethyl-, ethyl ester
116096
ethyl 4-propan-2-ylpiperazine-1-carboxylate
CCOC(=O)N1CCN(CC1)C(C)C
61014-91-3
C10H20N2O2
200.282
MFCD08064222
HRMZRPQIFGCXML-UHFFFAOYSA-N
ethyl 4-1-methylethyl piperazine-1-carboxylate,ethyl 4-isopropylpiperazine-1-carboxylate,ethyl 4-propan-2-yl piperazine-1-carboxylate,acmc-20akz8,ethyl 4-methylethyl piperazinecarboxylate,ethyl 4-isopropyl-1-piperazinecarboxylate,ethyl 4-1-methylethyl-1-piperazinylcarboxylate,4-isopropylpiperazine-1-carboxylic acid ethyl ester,1-piperazinecarboxylic acid, 4-1-methylethyl-, ethyl ester
116096
ethyl 4-propan-2-ylpiperazine-1-carboxylate
CCOC(=O)N1CCN(CC1)C(C)C
Quantity 1g
Boiling Point 128°C to 130°C (14mmHg)
Formula Weight 200.28
Percent Purity 98%
Chemical Name or Material Ethyl 4-isopropylpiperazine-1-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning