Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-methoxyphenylacetate, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
14062-18-1
C11H14O3
194.23
MFCD00040760
DOCCDOCIYYDLGJ-UHFFFAOYSA-N
ethyl 2-4-methoxyphenyl acetate,ethyl 4-methoxyphenylacetate,4-methoxyphenylacetic acid ethyl ester,benzeneacetic acid, 4-methoxy-, ethyl ester,ethyl p-methoxyphenylacetate,ethyl 4-methoxyphenyl acetate,4-methoxy-phenyl-acetic acid ethyl ester,acmc-1bnx2,homoanisic acid ethyl ester
84174
ethyl 2-(4-methoxyphenyl)acetate
CCOC(=O)CC1=CC=C(C=C1)OC
14062-18-1
C11H14O3
194.23
MFCD00040760
DOCCDOCIYYDLGJ-UHFFFAOYSA-N
ethyl 2-4-methoxyphenyl acetate,ethyl 4-methoxyphenylacetate,4-methoxyphenylacetic acid ethyl ester,benzeneacetic acid, 4-methoxy-, ethyl ester,ethyl p-methoxyphenylacetate,ethyl 4-methoxyphenyl acetate,4-methoxy-phenyl-acetic acid ethyl ester,acmc-1bnx2,homoanisic acid ethyl ester
84174
ethyl 2-(4-methoxyphenyl)acetate
CCOC(=O)CC1=CC=C(C=C1)OC
Refractive Index 1.5065 to 1.5085
Boiling Point 108°C to 111°C (3.0mmHg)
Specific Gravity 1.097
Flash Point 46°C
Quantity 10g
Density 1.0970g/mL
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula CH3OC6H4CH2CO2C2H5
Beilstein 10, I, 83
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material Ethyl 4-methoxyphenylacetate, 97%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 237-903-6