Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl 4-methyloxazole-5-carboxylate, 97%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
Specifications
Quantity | 1g |
Flash Point | 96°C |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Boiling Point | 100.0°C to 103.0°C (21.0 mmHg) |
Melting Point | 32.0°C to 38.0°C |
Assay Percent Range | 96% min. (GC) |
Formula Weight | 155.15 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 4-methyloxazole-5-carboxylate |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 243-848-9