Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-methyloxazole-5-carboxylate, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
20485-39-6
C7H9NO3
155.15
XNMORZSEENWFLI-UHFFFAOYSA-N
ethyl 4-methyloxazole-5-carboxylate,ethyl4-methyloxazole-5-carboxylate,4-methyloxazole-5-carboxylic acid ethyl ester,5-ethoxycarbonyl-4-methyloxazole,ethyl 4-methyl-5-oxazolecarboxylate,5-oxazolecarboxylic acid, 4-methyl-, ethyl ester,4-methyl-oxazole-5-carboxylic acid ethyl ester,4-methyl-5-carbethoxyoxazole,ethyl 4-methyl-oxazole-5-carboxylate,ethyl-4-methyl-1,3-oxazole-5-carboxylate
88558
ethyl 4-methyl-1,3-oxazole-5-carboxylate
CCOC(=O)C1=C(N=CO1)C
20485-39-6
C7H9NO3
155.15
XNMORZSEENWFLI-UHFFFAOYSA-N
ethyl 4-methyloxazole-5-carboxylate,ethyl4-methyloxazole-5-carboxylate,4-methyloxazole-5-carboxylic acid ethyl ester,5-ethoxycarbonyl-4-methyloxazole,ethyl 4-methyl-5-oxazolecarboxylate,5-oxazolecarboxylic acid, 4-methyl-, ethyl ester,4-methyl-oxazole-5-carboxylic acid ethyl ester,4-methyl-5-carbethoxyoxazole,ethyl 4-methyl-oxazole-5-carboxylate,ethyl-4-methyl-1,3-oxazole-5-carboxylate
88558
ethyl 4-methyl-1,3-oxazole-5-carboxylate
CCOC(=O)C1=C(N=CO1)C
Infrared Spectrum Authentic
Boiling Point 100.0°C to 103.0°C (21.0 mmHg)
Flash Point 96°C
Melting Point 32.0°C to 38.0°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 155.15
Percent Purity 97%
Chemical Name or Material Ethyl 4-methyloxazole-5-carboxylate

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 243-848-9