Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl 4-methyloxazole-5-carboxylate, 97+%, Thermo Scientific™
Click to view available options
Quantity:
250mg
1g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 250mg |
Formula Weight | 141.1 |
Percent Purity | ≥97% |
Chemical Name or Material | Ethyl 4-methyloxazole-5-carboxylate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 243-848-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only