Alfa Aesar™ Ethyl 4-nitrophenylacetate, 98%

Manufacturer: Alfa Aesar™ A1110122

Quantity 100g

 View more versions of this product

Catalog No. AAA1110122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5445-26-1
    Molecular Formula C10H11NO4
    Formula Weight 209.2
    MDL Number MFCD00017046
    Beilstein 2053241
    Synonym 4-Nitrophenylacetic acid ethyl ester
    Chemical Name or Material Ethyl 4-nitrophenylacetate

    Specifications

    Formula Weight 209.2
    MDL Number MFCD00017046
    Beilstein 2053241
    Quantity 100g
    TSCA TSCA
    Molecular Formula C10H11NO4
    CAS 5445-26-1
    Synonym 4-Nitrophenylacetic acid ethyl ester
    Melting Point 62° to 66°C
    Chemical Name or Material Ethyl 4-nitrophenylacetate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit