Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-nitrophenylacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
5445-26-1
C10H11NO4
209.201
DWDRNKYLWMKWTH-UHFFFAOYSA-N
ethyl 4-nitrophenylacetate,ethyl 2-4-nitrophenyl acetate,ethyl p-nitrophenylacetate,ethyl 4-nitrobenzeneacetate,ethyl 4-nitrophenyl acetate,4-nitrophenylacetic acid ethyl ester,benzeneacetic acid, 4-nitro-, ethyl ester,ethyl-4-nitrophenylacetate,acetic acid, p-nitrophenyl-, ethyl ester,4-nitro-phenyl-acetic acid ethyl ester
79517
ethyl 2-(4-nitrophenyl)acetate
CCOC(=O)CC1=CC=C(C=C1)[N+](=O)[O-]
5445-26-1
C10H11NO4
209.201
DWDRNKYLWMKWTH-UHFFFAOYSA-N
ethyl 4-nitrophenylacetate,ethyl 2-4-nitrophenyl acetate,ethyl p-nitrophenylacetate,ethyl 4-nitrobenzeneacetate,ethyl 4-nitrophenyl acetate,4-nitrophenylacetic acid ethyl ester,benzeneacetic acid, 4-nitro-, ethyl ester,ethyl-4-nitrophenylacetate,acetic acid, p-nitrophenyl-, ethyl ester,4-nitro-phenyl-acetic acid ethyl ester
79517
ethyl 2-(4-nitrophenyl)acetate
CCOC(=O)CC1=CC=C(C=C1)[N+](=O)[O-]
Quantity 5g
Infrared Spectrum Authentic
Color Yellow
Melting Point 61°C to 65°C
Assay Percent Range 98%
Linear Formula CH3CHOC(O)CHC6H4NO
Formula Weight 209.2
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Ethyl 4-nitrophenylacetate, 98%

EINECSNumber : 226-646-5

TSCA : TSCA