Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 4-pentenoate, 98+%

Manufacturer:  Alfa Aesar™ A1242222

Quantity 100g

 View more versions of this product

Catalog No. AAA1242222


  Check Availability
Add to cart

Specifications

Description

Applications
Ethyl 4-pentenoate is found in leaf oil of Prunus phaeosticta phaeosticta from Taiwan. It is also used in the preparation of 5-iodomethyl-dihydro-furan-2-one by reacting with iodine.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

1968-40-7
128.17
UN3272
Ethyl allylacetate; 4-Pentenoic acid ethyl ester
CH2=CHCH2CH2CO2CH2CH3
MFCD00036562
1751643
Ethyl 4-pentenoate

Specifications

37°C (98°F)
100g
CH2=CHCH2CH2CO2CH2CH3
MFCD00036562
1751643
Ethyl 4-pentenoate
146°C
1968-40-7
128.17
UN3272
Ethyl allylacetate; 4-Pentenoic acid ethyl ester
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit