Alfa Aesar™ Ethyl 5-bromo-2-chlorobenzoate, 98+%

Manufacturer: Alfa Aesar™ A1033414

Quantity 25g

 View more versions of this product

Catalog No. AAA1033414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76008-73-6
    Molecular Formula C9H8BrClO2
    Formula Weight 263.52
    MDL Number MFCD00013635
    Beilstein 6501991
    Synonym 5-Bromo-2-chlorobenzoic acid ethyl ester
    Chemical Name or Material Ethyl 5-bromo-2-chlorobenzoate

    Specifications

    Formula Weight 263.52
    Molecular Formula C9H8BrClO2
    Beilstein 6501991
    Synonym 5-Bromo-2-chlorobenzoic acid ethyl ester
    Boiling Point 156° to 157°C (13mmHg)
    Quantity 25g
    MDL Number MFCD00013635
    CAS 76008-73-6
    Flash Point >110°C (230°F)
    Chemical Name or Material Ethyl 5-bromo-2-chlorobenzoate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit