Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 5-bromoindole-2-carboxylate, 97%

Click to view available options
Quantity:
1g
5g
16732-70-0
C11H10BrNO2
268.11
MFCD00022701
LWRLKENDQISGEU-UHFFFAOYSA-N
5-bromoindole-2-carboxylic acid ethyl ester,ethyl 5-bromoindole-2-carboxylate,2-ethoxycarbonyl-5-bromo-indole,5-bromo-2-indolecarboxylic acid ethyl ester,5-bromo-1h-indole-2-carboxylic acid ethyl ester,5-bromoindole-2-carboxylicacidethylester,1h-indole-2-carboxylic acid, 5-bromo-, ethyl ester,5-bromo-2-carbethoxyindole,ethyl-5-brom-1h-indol-2-carboxylat,zlchem 465
259091
ethyl 5-bromo-1H-indole-2-carboxylate
CCOC(=O)C1=CC2=C(N1)C=CC(=C2)Br
16732-70-0
C11H10BrNO2
268.11
MFCD00022701
LWRLKENDQISGEU-UHFFFAOYSA-N
5-bromoindole-2-carboxylic acid ethyl ester,ethyl 5-bromoindole-2-carboxylate,2-ethoxycarbonyl-5-bromo-indole,5-bromo-2-indolecarboxylic acid ethyl ester,5-bromo-1h-indole-2-carboxylic acid ethyl ester,5-bromoindole-2-carboxylicacidethylester,1h-indole-2-carboxylic acid, 5-bromo-, ethyl ester,5-bromo-2-carbethoxyindole,ethyl-5-brom-1h-indol-2-carboxylat,zlchem 465
259091
ethyl 5-bromo-1H-indole-2-carboxylate
CCOC(=O)C1=CC2=C(N1)C=CC(=C2)Br
Melting Point 163°C to 167°C
Quantity 5g
Beilstein 170256
Formula Weight 268.11
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Ethyl 5-bromoindole-2-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C