Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 5-bromothiophene-2-carboxylate, 99%

Click to view available options
Quantity:
1g
5g
25g
5751-83-7
C7H7BrO2S
235.095
MFCD02683089
PZNHMXAOMDQLLE-UHFFFAOYSA-N
5-bromothiophene-2-carboxylic acid ethyl ester,ethyl 5-bromo-2-thiophenecarboxylate,ethyl5-bromothiophene-2-carboxylate,5-bromo-thiophene-2-carboxylic acid ethyl ester,2-thiophenecarboxylic acid, 5-bromo-, ethyl ester,pubchem16118,acmc-209zz5,ksc494s9b,2-bromo-5-ethoxycarbonylthiophene,ethyl 5-bromothiophene-2carboxylate
605723
ethyl 5-bromothiophene-2-carboxylate
CCOC(=O)C1=CC=C(S1)Br
5751-83-7
C7H7BrO2S
235.095
MFCD02683089
PZNHMXAOMDQLLE-UHFFFAOYSA-N
5-bromothiophene-2-carboxylic acid ethyl ester,ethyl 5-bromo-2-thiophenecarboxylate,ethyl5-bromothiophene-2-carboxylate,5-bromo-thiophene-2-carboxylic acid ethyl ester,2-thiophenecarboxylic acid, 5-bromo-, ethyl ester,pubchem16118,acmc-209zz5,ksc494s9b,2-bromo-5-ethoxycarbonylthiophene,ethyl 5-bromothiophene-2carboxylate
605723
ethyl 5-bromothiophene-2-carboxylate
CCOC(=O)C1=CC=C(S1)Br
Applications
Ethyl 5-bromothiophene-2-carboxylate is used as an intermediate in organic syntheses

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Quantity 25g
Boiling Point 94°C to 96°C (4mmHg)
Formula Weight 235.1
Percent Purity 99%
Chemical Name or Material Ethyl 5-bromothiophene-2-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning