Alfa Aesar™ Ethyl 5-fluorobenzo[b]thiophene-2-carboxylate, 96%

Manufacturer: Alfa Aesar™ H3416706

Quantity 5g

 View more versions of this product

Catalog No. AAH3416706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13771-69-2
    Molecular Formula C11H9FO2S
    Formula Weight 224.26
    MDL Number MFCD12923024
    Chemical Name or Material Ethyl 5-fluorobenzo[b]thiophene-2-carboxylate

    Specifications

    Formula Weight 224.26
    Molecular Formula C11H9FO2S
    MDL Number MFCD12923024
    CAS 13771-69-2
    Melting Point 88° to 89°C
    Quantity 5g
    Chemical Name or Material Ethyl 5-fluorobenzo[b]thiophene-2-carboxylate
    Assay 96%

    Description

    Notes
    Store at room temperature. Keep away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit