Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 5-fluorobenzo[b]thiophene-2-carboxylate, 96%

Manufacturer:  Alfa Aesar™ H34167MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH34167MD


  Check Availability
Add to cart

Specifications

Description

Notes
Store at room temperature. Keep away from oxidizing agents.
Description & Specifications

Chemical Identifiers

13771-69-2
224.26
Ethyl 5-fluorobenzo[b]thiophene-2-carboxylate
C11H9FO2S
MFCD12923024

Specifications

13771-69-2
224.26
88° to 89°C
Ethyl 5-fluorobenzo[b]thiophene-2-carboxylate
C11H9FO2S
MFCD12923024
250mg
96%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit