Alfa Aesar™ Ethyl 5-nitro-2-furoate, 97%

Manufacturer: Alfa Aesar™ B2486106

Quantity 5g

 View more versions of this product

Catalog No. AAB2486106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 943-37-3
    Molecular Formula C7H7NO5
    Formula Weight 185.14
    MDL Number MFCD00092566
    Synonym 5-Nitro-2-furoic acid ethyl ester
    Chemical Name or Material Ethyl 5-nitro-2-furoate

    Specifications

    Formula Weight 185.14
    Molecular Formula C7H7NO5
    RTECS Number LV2012500
    Quantity 5g
    MDL Number MFCD00092566
    CAS 943-37-3
    Synonym 5-Nitro-2-furoic acid ethyl ester
    Melting Point 98° to 101°C
    Chemical Name or Material Ethyl 5-nitro-2-furoate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit