Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl-6-(2-oxocyclopentyl)hexanoate 96%, Acros Organics

Click to view available options
Quantity:
500mg
63135-03-5
C13H22O3
226.316
MFCD00044716
LJZYHOVSZOHASC-UHFFFAOYSA-N
ethyl 6-2-oxocyclopentyl hexanoate,ethyl6-2-oxocyclopentyl hexanoate,6-2-oxocyclopentyl hexanoic acid ethyl ester,cyclopentanehexanoicacid, 2-oxo-, ethyl ester
2723664
ethyl 6-(2-oxocyclopentyl)hexanoate
CCOC(=O)CCCCCC1CCCC1=O
63135-03-5
C13H22O3
226.316
MFCD00044716
LJZYHOVSZOHASC-UHFFFAOYSA-N
ethyl 6-2-oxocyclopentyl hexanoate,ethyl6-2-oxocyclopentyl hexanoate,6-2-oxocyclopentyl hexanoic acid ethyl ester,cyclopentanehexanoicacid, 2-oxo-, ethyl ester
2723664
ethyl 6-(2-oxocyclopentyl)hexanoate
CCOC(=O)CCCCCC1CCCC1=O
Quantity 500mg
Infrared Spectrum Authentic
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Color Colorless
Assay Percent Range 96%
Formula Weight 226.31
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material Ethyl 6-(2-oxocyclopentyl)hexanoate, 96%